Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677130.3 | XP_005677187.1 | 1 - 2 | 262 | 95 | |
XM_013975375.2 | XP_013830829.1 | 1 - 2 | 259 | 95 | ||
GenBank mRNA | JO588627.2 | - | 3 - 4 | 481 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |