Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049023.1 | XP_017904512.1 | 3 - 4 | 649 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |