Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053051.1 | XP_017908540.1 | 3 - 4 | 657 | 95 | |
GenBank mRNA | JR114848.1 | - | 4 - 5 | 411 | 95 | |
JT632286.1 | - | 3 - 4 | 434 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |