Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685304.3 | XP_005685361.3 | 2 - 3 | 487 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |