Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695485.3 | XP_005695542.1 | 4 - 5 | 311 | 83 | |
XM_005695486.3 | XP_005695543.1 | 4 - 5 | 310 | 83 | ||
XM_018038281.1 | XP_017893770.1 | 4 - 5 | 311 | 83 | ||
XM_018038282.1 | XP_017893771.1 | 4 - 5 | 311 | 83 | ||
XM_018038283.1 | XP_017893772.1 | 4 - 5 | 311 | 83 | ||
XM_018038285.1 | XP_017893774.1 | 4 - 5 | 272 | 83 | ||
GenBank mRNA | JO605864.2 | - | 5 - 6 | 505 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |