Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677622.3 | XP_005677679.1 | 7 - 8 | 1544 | 61 | |
XM_005677623.3 | XP_005677680.1 | 7 - 8 | 869 | 61 | ||
GenBank mRNA | JO538851.2 | - | 7 - 8 | 893 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |