Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14H8orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689042.3 | XP_005689099.2 | 13 - 14 | 1816 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |