Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690007.3 | XP_005690064.1 | 14 - 15 | 2433 | 89 | |
XM_005690008.3 | XP_005690065.1 | 14 - 15 | 2432 | 89 | ||
XM_013969807.2 | XP_013825261.1 | 13 - 14 | 2391 | 89 | ||
XM_013969808.2 | XP_013825262.1 | 13 - 14 | 2280 | 89 | ||
XM_013969809.2 | XP_013825263.1 | 14 - 15 | 2434 | 89 | ||
XM_013969810.2 | XP_013825264.1 | 13 - 14 | 2279 | 89 | ||
XM_013969811.2 | XP_013825265.1 | 14 - 15 | 2432 | 89 | ||
XM_013969812.2 | XP_013825266.1 | 14 - 15 | 2434 | 89 | ||
XM_018059320.1 | XP_017914809.1 | 13 - 14 | 2333 | 89 | ||
GenBank mRNA | JT708455.1 | - | 12 - 13 | 2016 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |