Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061121.1 | XP_017916610.1 | 3 - 4 | 705 | 64 | |
GenBank mRNA | JO395985.2 | - | 3 - 4 | 265 | 64 | |
JO621595.2 | - | 3 - 4 | 482 | 64 | ||
KA314355.1 | - | 3 - 4 | 547 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |