Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693629.3 | XP_005693686.2 | 1 - 2 | 138 | 80 | |
XM_013972217.2 | XP_013827671.1 | 2 - 3 | 306 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |