Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFRP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698387.3 | XP_005698444.3 | 1 - 2 | 575 | 101 | |
GenBank mRNA | JT671586.1 | - | 3 | 604 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Capra hircus Probe spans exons |