Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699520.3 | XP_005699577.1 | 2 - 3 | 270 | 57 | |
GenBank mRNA | JP350874.2 | - | 4 - 5 | 668 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |