Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058360.1 | XP_017913849.1 | 2 - 3 | 315 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |