Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682502.2 | XP_005682559.1 | 2 - 3 | 1501 | 80 | |
GenBank mRNA | KA324844.1 | - | 2 - 3 | 766 | 80 |
Gene Symbol: RANBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO646009.2 | - | 18 - 19 | 3746 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |