Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686638.3 | XP_005686695.2 | 5 - 6 | 683 | 73 | |
GenBank mRNA | JT680139.1 | - | 3 - 4 | 729 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Endogenous Controls Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |