Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM72A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690410.3 | XP_005690467.1 | 2 - 3 | 1115 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |