Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM72A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690410.3 | XP_005690467.1 | 3 - 4 | 1242 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |