Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695348.3 | XP_005695405.1 | 4 - 5 | 1273 | 82 | |
XM_005695349.3 | XP_005695406.1 | 4 - 5 | 1253 | 82 | ||
GenBank mRNA | JO420012.2 | - | 4 - 5 | 1274 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |