Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698497.3 | XP_005698554.1 | 5 - 6 | 690 | 68 | |
XM_018041622.1 | XP_017897111.1 | 5 - 6 | 574 | 68 | ||
GenBank mRNA | JI861721.1 | - | 4 - 5 | 559 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |