Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEANC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043867.1 | XP_017899356.1 | 2 - 3 | 192 | 72 | |
XM_018043868.1 | XP_017899357.1 | 2 - 3 | 162 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |