Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013969062.2 | XP_013824516.2 | 2 - 3 | 2322 | 80 |
Gene Symbol: GLI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO509821.2 | - | 2 - 3 | 770 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |