Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039810.1 | XP_017895299.1 | 4 - 5 | 868 | 68 | |
XM_018039811.1 | XP_017895300.1 | 2 - 3 | 606 | 68 | ||
GenBank mRNA | JO623001.2 | - | 1024 | 68 | ||
JO623007.2 | - | 4 - 5 | 887 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |