Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679316.3 | XP_005679373.1 | 8 - 9 | 1723 | 58 | |
XM_013963385.2 | XP_013818839.1 | 8 - 9 | 1640 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |