Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IZUMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683598.3 | XP_005683655.1 | 3 - 4 | 629 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |