Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681312.3 | XP_005681369.2 | 3 - 4 | 674 | 99 | |
XM_005681313.2 | XP_005681370.2 | 3 - 4 | 467 | 99 | ||
GenBank mRNA | JT653501.1 | - | 5 - 6 | 478 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |