Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687432.3 | XP_005687489.1 | 9 - 10 | 1427 | 91 | |
XM_018056765.1 | XP_017912254.1 | 9 - 10 | 1011 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |