Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680910.3 | XP_005680967.2 | 2 - 3 | 367 | 84 | |
GenBank mRNA | JO335635.2 | - | 4 - 5 | 628 | 84 | |
JO590103.2 | - | 2 - 3 | 422 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |