Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681322.2 | XP_005681379.1 | 1 - 2 | 206 | 70 | |
GenBank mRNA | JO412291.2 | - | 1 - 2 | 217 | 70 |
Gene Symbol: LOC108634094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042119.1 | XP_017897608.1 | 1 | 196 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |