Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057690.1 | XP_017913179.1 | 10 - 11 | 1325 | 70 | |
GenBank mRNA | JO610655.2 | - | 932 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |