Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053511.1 | XP_017909000.1 | 2 - 3 | 225 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Capra hircus Probe spans exons |