Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043289.1 | XP_017898778.1 | 3 - 4 | 1567 | 77 | |
XM_018043292.1 | XP_017898781.1 | 3 - 4 | 1458 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |