Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043289.1 | XP_017898778.1 | 5 - 6 | 2036 | 77 | |
XM_018043290.1 | XP_017898779.1 | 5 - 6 | 1986 | 77 | ||
XM_018043291.1 | XP_017898780.1 | 5 - 6 | 1982 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |