Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054870.1 | XP_017910359.1 | 3 - 4 | 471 | 59 | |
GenBank mRNA | JO438628.2 | - | 3 - 4 | 431 | 59 | |
JO640088.2 | - | 3 - 4 | 451 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |