Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059818.1 | XP_017915307.1 | 5 - 6 | 1393 | 101 | |
XM_018059819.1 | XP_017915308.1 | 5 - 6 | 1288 | 101 | ||
GenBank mRNA | JT743132.1 | - | 4 - 5 | 1248 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |