Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680675.3 | XP_005680732.1 | 2 - 3 | 413 | 83 | |
GenBank mRNA | JO357419.2 | - | 1 - 2 | 2323 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |