Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSPRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052628.1 | XP_017908117.1 | 5 - 6 | 727 | 70 | |
XM_018052629.1 | XP_017908118.1 | 5 - 6 | 725 | 70 | ||
GenBank mRNA | KA331230.1 | - | 5 - 6 | 687 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |