Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054988.1 | XP_017910477.1 | 2 - 3 | 972 | 87 | |
GenBank mRNA | JO577843.1 | - | 2 - 3 | 447 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |