Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690840.2 | XP_005690897.2 | 10 - 11 | 1423 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |