Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041479.1 | XP_017896968.1 | 2 - 3 | 1243 | 69 | |
XM_018041480.1 | XP_017896969.1 | 4 - 5 | 1468 | 69 | ||
XM_018041481.1 | XP_017896970.1 | 3 - 4 | 1326 | 69 | ||
XM_018041482.1 | XP_017896971.1 | 2 - 3 | 1120 | 69 | ||
XM_018041483.1 | XP_017896972.1 | 3 - 4 | 1385 | 69 | ||
XM_018041484.1 | XP_017896973.1 | 2 - 3 | 1519 | 69 | ||
GenBank mRNA | JO426060.1 | - | 4 - 5 | 1522 | 69 | |
JO605079.2 | - | 1070 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |