Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039462.1 | XP_017894951.1 | 5 - 6 | 4447 | 85 | |
XM_018039463.1 | XP_017894952.1 | 5 - 6 | 4455 | 85 | ||
XM_018039464.1 | XP_017894953.1 | 4 - 5 | 4339 | 85 | ||
XM_018039465.1 | XP_017894954.1 | 3 - 4 | 4285 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |