Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681841.3 | XP_005681898.2 | 2 - 3 | 424 | 83 | |
XM_005681842.3 | XP_005681899.2 | 2 - 3 | 325 | 83 | ||
XM_013964756.2 | XP_013820210.2 | 2 - 3 | 416 | 83 | ||
GenBank mRNA | JO594467.2 | - | 2 - 3 | 411 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |