Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038665.1 | XP_017894154.1 | 3 - 4 | 412 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |