Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675326.3 | XP_005675383.2 | 1 - 2 | 511 | 74 | |
GenBank mRNA | JO633118.2 | - | 1 - 2 | 451 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |