Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055430.1 | XP_017910919.1 | 6 - 7 | 1316 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |