Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL21C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687731.3 | XP_005687788.2 | 3 - 4 | 915 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |