Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042082.1 | XP_017897571.1 | 5 - 6 | 1074 | 84 | |
GenBank mRNA | JO567232.2 | - | 2 - 3 | 170 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |