Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO633415.2 | - | 904 | 70 |
Gene Symbol: SYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060849.1 | XP_017916338.1 | 5 - 6 | 814 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |