Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039259.1 | XP_017894748.1 | 2 - 3 | 873 | 61 | |
XM_018039260.1 | XP_017894749.1 | 2 - 3 | 320 | 61 | ||
GenBank mRNA | JO348333.2 | - | 2 - 3 | 386 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |