Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680868.2 | XP_005680925.1 | 4 - 5 | 569 | 96 | |
GenBank mRNA | JO605363.2 | - | 4 - 5 | 623 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |