Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687692.3 | XP_005687749.2 | 5 - 6 | 1254 | 65 | |
XM_018056508.1 | XP_017911997.1 | 5 - 6 | 1299 | 65 | ||
GenBank mRNA | KX247669.1 | - | 5 - 6 | 859 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |